Product Details

SNP ID
rs114428333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102943638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGTCATTGCCAAAAGTGAAAAT[C/T]AAACAGAATTCAAAGGCCACATCTA
Phenotype
MIM: 600108
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMP13 PubMed Links

Gene Details

Gene
MMP13
Gene Name
matrix metallopeptidase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002427.3 2073 UTR 3 NP_002418.1

View Full Product Details