Product Details

SNP ID
rs137997569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103967800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTTGGGGAGCGAGAAGAAGAAG[A/C]AGCAGTACGAACACGTGAAGAGGGA
Phenotype
MIM: 616563
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLK PubMed Links

Gene Details

Gene
SLK
Gene Name
STE20 like kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304743.1 628 Missense Mutation AAG,CAG K19Q NP_001291672.1
NM_014720.3 628 Missense Mutation AAG,CAG K19Q NP_055535.2
XM_011540401.2 628 Missense Mutation AAG,CAG K19Q XP_011538703.1

View Full Product Details