Product Details
- SNP ID
-
rs138995637
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:59247737 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTATCCTATCTTCCTTTTGTGGACT[C/T]CTGCAAAACAAAAATACATTTGTTA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM13C
PubMed Links
Gene Details
- Gene
- FAM13C
- Gene Name
- family with sequence similarity 13 member C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001971.2 |
1952 |
Silent Mutation |
AGA,AGG |
R447R |
NP_001001971.1 |
NM_001143773.1 |
1952 |
Silent Mutation |
AGA,AGG |
R462R |
NP_001137245.1 |
NM_001166698.1 |
1952 |
Silent Mutation |
AGA,AGG |
R461R |
NP_001160170.1 |
NM_198215.3 |
1952 |
Silent Mutation |
AGA,AGG |
R545R |
NP_937858.2 |
XM_005269618.4 |
1952 |
Silent Mutation |
AGA,AGG |
R544R |
XP_005269675.2 |
XM_005269619.4 |
1952 |
Silent Mutation |
AGA,AGG |
R446R |
XP_005269676.2 |
XM_006717702.3 |
1952 |
Silent Mutation |
AGA,AGG |
R462R |
XP_006717765.1 |
XM_006717703.3 |
1952 |
Silent Mutation |
AGA,AGG |
R462R |
XP_006717766.1 |
XM_011539490.2 |
1952 |
Silent Mutation |
AGA,AGG |
R453R |
XP_011537792.1 |
XM_011539491.2 |
1952 |
Silent Mutation |
AGA,AGG |
R453R |
XP_011537793.1 |
XM_017015885.1 |
1952 |
Silent Mutation |
AGA,AGG |
R462R |
XP_016871374.1 |
XM_017015886.1 |
1952 |
Silent Mutation |
AGA,AGG |
R461R |
XP_016871375.1 |
XM_017015887.1 |
1952 |
Silent Mutation |
AGA,AGG |
R461R |
XP_016871376.1 |
XM_017015888.1 |
1952 |
Silent Mutation |
AGA,AGG |
R461R |
XP_016871377.1 |
XM_017015889.1 |
1952 |
Silent Mutation |
AGA,AGG |
R461R |
XP_016871378.1 |
XM_017015890.1 |
1952 |
Silent Mutation |
AGA,AGG |
R452R |
XP_016871379.1 |
XM_017015891.1 |
1952 |
Silent Mutation |
AGA,AGG |
R364R |
XP_016871380.1 |
XM_017015892.1 |
1952 |
Silent Mutation |
AGA,AGG |
R363R |
XP_016871381.1 |
XM_017015893.1 |
1952 |
Silent Mutation |
AGA,AGG |
R355R |
XP_016871382.1 |
- Gene
- PHYHIPL
- Gene Name
- phytanoyl-CoA 2-hydroxylase interacting protein like
View Full Product Details