Product Details

SNP ID
rs139051724
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:16828840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGTCCAGAGGATGGAGAGCTGG[C/T]GTTGGGCCCATCATAGAGTGTGAGA
Phenotype
MIM: 602997
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CUBN PubMed Links

Gene Details

Gene
CUBN
Gene Name
cubilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081.3 7144 Missense Mutation ACC,GCC T3577A NP_001072.2
XM_011519708.2 7144 Intron XP_011518010.1
XM_011519709.2 7144 Missense Mutation ACC,GCC T2239A XP_011518011.1
XM_011519710.2 7144 Missense Mutation ACC,GCC T2231A XP_011518012.1
XM_011519711.2 7144 Missense Mutation ACC,GCC T2191A XP_011518013.1

View Full Product Details