Product Details

SNP ID
rs145650735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:651457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCGTTAAGTAAAAATAGCAGAAG[A/T]CTTAGTAGAATGTATGAGTAACAAT
Phenotype
MIM: 611380
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DIP2C PubMed Links

Gene Details

Gene
DIP2C
Gene Name
disco interacting protein 2 homolog C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014974.2 Intron NP_055789.1
XM_005252426.3 Intron XP_005252483.1
XM_005252427.3 Intron XP_005252484.1
XM_005252428.4 Intron XP_005252485.1
XM_005252430.2 Intron XP_005252487.2
XM_011519428.2 Intron XP_011517730.1
XM_011519429.2 Intron XP_011517731.1
XM_011519430.2 Intron XP_011517732.1
XM_011519431.2 Intron XP_011517733.1
XM_011519432.1 Intron XP_011517734.1
XM_011519433.2 Intron XP_011517735.1
XM_017015977.1 Intron XP_016871466.1
Gene
MIR5699
Gene Name
microRNA 5699
There are no transcripts associated with this gene.

Gene
PRR26
Gene Name
proline rich 26
There are no transcripts associated with this gene.

View Full Product Details