Product Details

SNP ID
rs151134377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:16825013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAATCGGAATGCGGATGGACGCC[A/G]TGCATAATCAGCATGAAATTTTATG
Phenotype
MIM: 602997 MIM: 179555
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CUBN PubMed Links

Gene Details

Gene
CUBN
Gene Name
cubilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081.3 7249 Missense Mutation CGG,TGG R3612W NP_001072.2
XM_011519708.2 7249 Intron XP_011518010.1
XM_011519709.2 7249 Missense Mutation CGG,TGG R2274W XP_011518011.1
XM_011519710.2 7249 Missense Mutation CGG,TGG R2266W XP_011518012.1
XM_011519711.2 7249 Missense Mutation CGG,TGG R2226W XP_011518013.1
Gene
RSU1
Gene Name
Ras suppressor protein 1
There are no transcripts associated with this gene.

View Full Product Details