Product Details

SNP ID
rs115615172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61901911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGAACTGCGCTATTTTAAGCTA[A/C]CAGGTTGATAGTAATTTGTTACACC
Phenotype
MIM: 606150
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FADS3 PubMed Links

Gene Details

Gene
FADS3
Gene Name
fatty acid desaturase 3
There are no transcripts associated with this gene.

Gene
RAB3IL1
Gene Name
RAB3A interacting protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271686.1 Intron NP_001258615.1
NM_013401.3 Intron NP_037533.2
XM_005274140.4 Intron XP_005274197.1
XM_006718647.3 Intron XP_006718710.1
XM_011545196.2 Intron XP_011543498.1
XM_011545197.2 Intron XP_011543499.1
XM_011545198.2 Intron XP_011543500.1
XM_011545199.2 Intron XP_011543501.1
XM_011545200.2 Intron XP_011543502.1
XM_011545201.2 Intron XP_011543503.1

View Full Product Details