Product Details

SNP ID
rs117118339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57464230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAATGTGGGCAGGTGGGGAGCAGG[A/G]GAGTCTAGGGAGAGAAAGAGGAGTA
Phenotype
MIM: 610462
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RTN4RL2 PubMed Links

Gene Details

Gene
RTN4RL2
Gene Name
reticulon 4 receptor-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178570.2 Intron NP_848665.1

View Full Product Details