Product Details

SNP ID
rs138580044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:9142021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATTGTTGATTGCAGTGACAAAT[C/T]GGCAGAAGTTCCGGGCTCTTGTATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DENND5A PubMed Links

Gene Details

Gene
DENND5A
Gene Name
DENN domain containing 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243254.1 2119 Missense Mutation CAA,CGA Q1200R NP_001230183.1
NM_015213.3 2119 Missense Mutation CAA,CGA Q1200R NP_056028.2
XM_005252832.1 2119 Missense Mutation CAA,CGA Q1200R XP_005252889.1
XM_011519953.2 2119 Missense Mutation CAA,CGA Q566R XP_011518255.1
XM_017017419.1 2119 Intron XP_016872908.1

View Full Product Details