Product Details

SNP ID
rs138657153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64317139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCGGCCCTTTCGGCACCTGGAT[C/G]AGACGCAACTGTGGGCAAGAGGCCA
Phenotype
MIM: 601998 MIM: 606583
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ESRRA PubMed Links

Gene Details

Gene
ESRRA
Gene Name
estrogen related receptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282450.1 747 Intron NP_001269379.1
NM_001282451.1 747 Intron NP_001269380.1
NM_004451.4 747 Intron NP_004442.3
XM_017017313.1 747 Intron XP_016872802.1
Gene
PRDX5
Gene Name
peroxiredoxin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012094.4 747 Intron NP_036226.1
NM_181651.2 747 Intron NP_857634.1
NM_181652.2 747 Intron NP_857635.1
XM_005273882.4 747 Intron XP_005273939.2
Gene
TRMT112
Gene Name
tRNA methyltransferase 11-2 homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286082.1 747 Intron NP_001273011.1
NM_001286084.1 747 Silent Mutation CTC,CTG L63L NP_001273013.1
NM_016404.2 747 Silent Mutation CTC,CTG L63L NP_057488.1

View Full Product Details