Product Details

SNP ID
rs139324033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124885077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATACGACAGTGGAGCTGACTTC[A/T]CTGGGAAGAGATCTGAGAGTCAGGG
Phenotype
MIM: 608630 MIM: 607528
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ROBO3 PubMed Links

Gene Details

Gene
ROBO3
Gene Name
roundabout guidance receptor 3
There are no transcripts associated with this gene.

Gene
ROBO4
Gene Name
roundabout guidance receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301088.1 3105 Nonsense Mutation AGA,TGA R844* NP_001288017.1
NM_019055.5 3105 Nonsense Mutation AGA,TGA R989* NP_061928.4
XM_006718861.1 3105 Nonsense Mutation AGA,TGA R951* XP_006718924.1
XM_011542875.1 3105 Nonsense Mutation AGA,TGA R547* XP_011541177.1

View Full Product Details