Product Details

SNP ID
rs139612286
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:33703963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGTACCTGAGGTGCTGTTTGCC[A/G]GGTTCCCTTGGTCCTCAGACTGCTC
Phenotype
MIM: 107271
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C11orf91 PubMed Links

Gene Details

Gene
C11orf91
Gene Name
chromosome 11 open reading frame 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166692.1 6725 Intron NP_001160164.1
XM_017017053.1 6725 Intron XP_016872542.1
Gene
CD59
Gene Name
CD59 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000611.5 6725 UTR 3 NP_000602.1
NM_001127223.1 6725 UTR 3 NP_001120695.1
NM_001127225.1 6725 UTR 3 NP_001120697.1
NM_001127226.1 6725 UTR 3 NP_001120698.1
NM_001127227.1 6725 UTR 3 NP_001120699.1
NM_203329.2 6725 UTR 3 NP_976074.1
NM_203330.2 6725 UTR 3 NP_976075.1
NM_203331.2 6725 UTR 3 NP_976076.1

View Full Product Details