Product Details

SNP ID
rs139731858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:9142083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGACTACTTCATTCTTCTCTAA[C/T]GTCTCATAATAGGTTTGTGCTTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DENND5A PubMed Links

Gene Details

Gene
DENND5A
Gene Name
DENN domain containing 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243254.1 2057 Silent Mutation ACA,ACG T1179T NP_001230183.1
NM_015213.3 2057 Silent Mutation ACA,ACG T1179T NP_056028.2
XM_005252832.1 2057 Silent Mutation ACA,ACG T1179T XP_005252889.1
XM_011519953.2 2057 Silent Mutation ACA,ACG T545T XP_011518255.1
XM_017017419.1 2057 Intron XP_016872908.1

View Full Product Details