Product Details

SNP ID
rs140636199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7825431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGCAAGCTTCAAAAGTGGTGAA[C/T]AGTCACAGAAAAAGTGATTGACTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5P3 PubMed Links

Gene Details

Gene
OR5P3
Gene Name
olfactory receptor family 5 subfamily P member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153445.1 542 Missense Mutation TAT,TGT Y181C NP_703146.1

View Full Product Details