Product Details

SNP ID
rs141267245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6108345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACTGCCTCTGCTCTAACTTGGG[G/T]GTTATCAGCCTGGCTTGTGATGACA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR56B4 PubMed Links

Gene Details

Gene
OR56B4
Gene Name
olfactory receptor family 56 subfamily B member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005181.2 662 Silent Mutation GGG,GGT G189G NP_001005181.1

View Full Product Details