Product Details

SNP ID
rs141714712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4587780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCACCTTAAATCCCATCATCT[A/G]TGGCATGAGGACCAAACAACTGCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf40 PubMed Links

Gene Details

Gene
C11orf40
Gene Name
chromosome 11 open reading frame 40
There are no transcripts associated with this gene.

Gene
OR52I1
Gene Name
olfactory receptor family 52 subfamily I member 1
There are no transcripts associated with this gene.

Gene
OR52I2
Gene Name
olfactory receptor family 52 subfamily I member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005170.2 990 Missense Mutation TAT,TGT Y323C NP_001005170.1

View Full Product Details