Product Details
- SNP ID
-
rs142596370
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:124886505 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCTCCCTGGGCTCTAGACCGAAA[C/T]CAAAGCTATCCACAGCCACTGCCAG
- Phenotype
-
MIM: 608630
MIM: 607528
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ROBO3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7104934] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ROBO3
- Gene Name
- roundabout guidance receptor 3
There are no transcripts associated with this gene.
- Gene
- ROBO4
- Gene Name
- roundabout guidance receptor 4
View Full Product Details