Product Details
- SNP ID
-
rs143804085
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:74764148 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGCAATAATCAGACTGAGAGCAT[A/G]TATCTTTTAACCTACAAGAGAGACC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNF169
PubMed Links
Gene Details
- Gene
- RNF169
- Gene Name
- ring finger protein 169
View Full Product Details