Product Details

SNP ID
rs145780180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66362826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGTGGGTATGGAAGAGCTTTG[C/T]AAACTGTAAAGTGCAGTACACACAT
Phenotype
MIM: 602110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724064 PubMed Links

Gene Details

Gene
LOC102724064
Gene Name
uncharacterized LOC102724064
There are no transcripts associated with this gene.

Gene
SLC29A2
Gene Name
solute carrier family 29 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300868.1 2361 UTR 3 NP_001287797.1
NM_001300869.1 2361 UTR 3 NP_001287798.1
NM_001532.2 2361 UTR 3 NP_001523.2
XM_011544974.1 2361 UTR 3 XP_011543276.1
XM_017017630.1 2361 UTR 3 XP_016873119.1
XM_017017631.1 2361 Intron XP_016873120.1
XM_017017632.1 2361 UTR 3 XP_016873121.1
XM_017017633.1 2361 Intron XP_016873122.1
XM_017017634.1 2361 Intron XP_016873123.1
XM_017017635.1 2361 UTR 3 XP_016873124.1
XM_017017636.1 2361 Intron XP_016873125.1
XM_017017637.1 2361 UTR 3 XP_016873126.1
XM_017017638.1 2361 UTR 3 XP_016873127.1

View Full Product Details