Product Details

SNP ID
rs146230377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124885058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTCACCACCAGCCTTGGGCATA[C/T]GACAGTGGAGCTGACTTCTCTGGGA
Phenotype
MIM: 608630 MIM: 607528
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ROBO3 PubMed Links

Gene Details

Gene
ROBO3
Gene Name
roundabout guidance receptor 3
There are no transcripts associated with this gene.

Gene
ROBO4
Gene Name
roundabout guidance receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301088.1 3124 Missense Mutation CAT,CGT H850R NP_001288017.1
NM_019055.5 3124 Missense Mutation CAT,CGT H995R NP_061928.4
XM_006718861.1 3124 Missense Mutation CAT,CGT H957R XP_006718924.1
XM_011542875.1 3124 Missense Mutation CAT,CGT H553R XP_011541177.1

View Full Product Details