Product Details

SNP ID
rs146811425
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57467742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCAACAACTTCTCCTCTGTGCC[A/G]CTGTCCCTGCCACCCAGCACTCAGC
Phenotype
MIM: 610462
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RTN4RL2 PubMed Links

Gene Details

Gene
RTN4RL2
Gene Name
reticulon 4 receptor-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178570.2 494 Silent Mutation CCA,CCG P55P NP_848665.1

View Full Product Details