Product Details

SNP ID
rs147182065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113780054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGCCCGCATCCTCATGGTAGCC[A/T]CCCATGGGCTGGGCCTATTGGGGCT
Phenotype
MIM: 603954
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CLDN25 PubMed Links

Gene Details

Gene
CLDN25
Gene Name
claudin 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101389.1 259 Missense Mutation ACC,TCC T87S NP_001094859.1
Gene
LOC390251
Gene Name
SH3 domain containing GRB2 like 1 pseudogene
There are no transcripts associated with this gene.

Gene
ZW10
Gene Name
zw10 kinetochore protein
There are no transcripts associated with this gene.

View Full Product Details