Product Details

SNP ID
rs147267631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61397880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATTACCTGCTGCTGCAGACCTA[C/T]GTACTCCGCCTGGAAGCCATCATGA
Phenotype
MIM: 613277
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPSF7 PubMed Links

Gene Details

Gene
CPSF7
Gene Name
cleavage and polyadenylation specific factor 7
There are no transcripts associated with this gene.

Gene
TMEM216
Gene Name
transmembrane protein 216
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173990.2 1295 Silent Mutation TAC,TAT Y112Y NP_001167461.1
NM_001173991.2 1295 Silent Mutation TAC,TAT Y112Y NP_001167462.1
NM_016499.5 1295 Silent Mutation TAC,TAT Y51Y NP_057583.2
XM_005274039.3 1295 Silent Mutation TAC,TAT Y51Y XP_005274096.1
XM_017017882.1 1295 Silent Mutation TAC,TAT Y51Y XP_016873371.1

View Full Product Details