Product Details

SNP ID
rs150568011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4587852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCATGTCCTCTTTGACCATTCCA[A/T]CCTGGGTTCATGAACACAATATCTG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR52I1 PubMed Links

Gene Details

Gene
OR52I1
Gene Name
olfactory receptor family 52 subfamily I member 1
There are no transcripts associated with this gene.

Gene
OR52I2
Gene Name
olfactory receptor family 52 subfamily I member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005170.2 1062 Missense Mutation AAC,ATC N347I NP_001005170.1

View Full Product Details