Product Details

SNP ID
rs150793793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58358453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCAAGGTAACAAGAAGTGCAAAA[A/C]AGACATTAAAACTTGATATAAGAAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR5B17 PubMed Links

Gene Details

Gene
OR5B17
Gene Name
olfactory receptor family 5 subfamily B member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005489.1 617 Missense Mutation TGT,TTT C206F NP_001005489.1

View Full Product Details