Product Details

SNP ID
rs151257734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:32587013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATAATAGAGCAATCCTCAACAGG[C/G]TGCTGAGCTTCGTGCTTATCTGAAA
Phenotype
MIM: 609641
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EIF3M PubMed Links

Gene Details

Gene
EIF3M
Gene Name
eukaryotic translation initiation factor 3 subunit M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307929.1 189 Intron NP_001294858.1
NM_006360.5 189 Missense Mutation GCT,GGT A15G NP_006351.2
XM_011519840.2 189 UTR 5 XP_011518142.1

View Full Product Details