Product Details

SNP ID
rs139405986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56207717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTTGACACTGCGGATTGCACG[A/C]ATGTATGCCTTTAGCAGCTGGATGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RNF41 PubMed Links

Gene Details

Gene
RNF41
Gene Name
ring finger protein 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242826.1 962 Missense Mutation ATG,ATT M177I NP_001229755.1
NM_005785.3 962 Missense Mutation ATG,ATT M177I NP_005776.1
NM_194358.2 962 Missense Mutation ATG,ATT M106I NP_919339.1
NM_194359.2 962 Missense Mutation ATG,ATT M177I NP_919340.1
XM_005268561.4 962 Missense Mutation ATG,ATT M106I XP_005268618.1
XM_011537734.2 962 Missense Mutation ATG,ATT M106I XP_011536036.1
XM_011537735.2 962 Missense Mutation ATG,ATT M106I XP_011536037.1

View Full Product Details