Product Details

SNP ID
rs140220052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:96001014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATCCACTTTTAAGTCTTTCCCC[A/G]CCAGCATTGCAGTCACGGGATGCAT
Phenotype
MIM: 609457 MIM: 151570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAL PubMed Links

Gene Details

Gene
HAL
Gene Name
histidine ammonia-lyase
There are no transcripts associated with this gene.

Gene
LTA4H
Gene Name
leukotriene A4 hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000895.2 1860 Missense Mutation GCG,GTG A604V NP_000886.1
NM_001256643.1 1860 Missense Mutation GCG,GTG A580V NP_001243572.1
NM_001256644.1 1860 UTR 3 NP_001243573.1
XM_005268871.1 1860 UTR 3 XP_005268928.1
XM_011538348.1 1860 Intron XP_011536650.1
XM_011538349.2 1860 Intron XP_011536651.1

View Full Product Details