Product Details

SNP ID
rs140608017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123458166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGGCGCGGAGAATCTGCTGTCG[C/T]CTGCAGCTGCTCGCCTGTCTCCGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNRNP35 PubMed Links

Gene Details

Gene
SNRNP35
Gene Name
small nuclear ribonucleoprotein U11/U12 subunit 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022717.3 523 UTR 5 NP_073208.1
NM_180699.3 523 Intron NP_851030.1
XM_006719204.3 523 Intron XP_006719267.1
XM_011537780.2 523 Missense Mutation CCT,TCT P44S XP_011536082.1
XM_017018724.1 523 UTR 5 XP_016874213.1

View Full Product Details