Product Details

SNP ID
rs142121934
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55400747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTTAGAAATTTCATTTACGAC[A/G]GTCTTCATTCCTAGATTTCTGATCA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR6C65 PubMed Links

Gene Details

Gene
OR6C65
Gene Name
olfactory receptor family 6 subfamily C member 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005518.1 219 Silent Mutation ACA,ACG T73T NP_001005518.1

View Full Product Details