Product Details

SNP ID
rs145461300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55400758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCATTTACGACAGTCTTCATTC[C/G]TAGATTTCTGATCAACATTGCTACA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR6C65 PubMed Links

Gene Details

Gene
OR6C65
Gene Name
olfactory receptor family 6 subfamily C member 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005518.1 230 Missense Mutation CCT,CGT P77R NP_001005518.1

View Full Product Details