Product Details

SNP ID
rs145862157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:124947180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCGGTGACCAAGCACAGCCAAC[A/G]TCAGCTCCGCTCCCTGCCGTCTGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHX37 PubMed Links

Gene Details

Gene
DHX37
Gene Name
DEAH-box helicase 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032656.3 4195 UTR 3 NP_116045.2
XM_005253590.3 4195 Intron XP_005253647.1
XM_011538598.2 4195 Intron XP_011536900.1
XM_011538600.2 4195 Intron XP_011536902.1

View Full Product Details