Product Details

SNP ID
rs146208493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123465621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATGGCACCGATGAAGACCCACA[C/T]GACCGCGCGGTCTGGAGGGCAATGC
Phenotype
MIM: 614092
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RILPL1 PubMed Links

Gene Details

Gene
RILPL1
Gene Name
Rab interacting lysosomal protein like 1
There are no transcripts associated with this gene.

Gene
SNRNP35
Gene Name
small nuclear ribonucleoprotein U11/U12 subunit 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022717.3 1380 Silent Mutation CAC,CAT H27H NP_073208.1
NM_180699.3 1380 Silent Mutation CAC,CAT H32H NP_851030.1
XM_006719204.3 1380 Silent Mutation CAC,CAT H27H XP_006719267.1
XM_011537780.2 1380 Silent Mutation CAC,CAT H88H XP_011536082.1
XM_017018724.1 1380 Silent Mutation CAC,CAT H32H XP_016874213.1

View Full Product Details