Product Details

SNP ID
rs146265569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:31382343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATATGTATGTTAAAGTGCTTAAT[A/G]TCACAATACTCTTTCAAATTTTATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DENND5B PubMed Links

Gene Details

Gene
DENND5B
Gene Name
DENN domain containing 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308339.1 9384 UTR 3 NP_001295268.1
NM_144973.3 9384 UTR 3 NP_659410.3
XM_005253326.2 9384 UTR 3 XP_005253383.1
XM_005253327.3 9384 Intron XP_005253384.1
XM_005253330.3 9384 Intron XP_005253387.1
XM_006719040.3 9384 Intron XP_006719103.1
XM_011520590.2 9384 Intron XP_011518892.1
XM_017018896.1 9384 UTR 3 XP_016874385.1
XM_017018897.1 9384 Intron XP_016874386.1
XM_017018898.1 9384 Intron XP_016874387.1
XM_017018899.1 9384 Intron XP_016874388.1

View Full Product Details