Product Details

SNP ID
rs148263008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55401186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGTTCTCTCCTACACACTCATC[C/T]TTAAAACAATTCTGAAGATTCCCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6C65 PubMed Links

Gene Details

Gene
OR6C65
Gene Name
olfactory receptor family 6 subfamily C member 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005518.1 658 Missense Mutation CTT,TTT L220F NP_001005518.1

View Full Product Details