Product Details
- SNP ID
-
rs148661122
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:118160251 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGTGTCTTTAAGATTGTTTTGTG[A/C]TCATTCTTTGGAGTAACTTCCAACT
- Phenotype
-
MIM: 616711
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TAOK3
PubMed Links
Gene Details
- Gene
- TAOK3
- Gene Name
- TAO kinase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016281.3 |
3071 |
Missense Mutation |
GAG,GAT |
E749D |
NP_057365.3 |
XM_005253897.1 |
3071 |
Missense Mutation |
GAG,GAT |
E758D |
XP_005253954.1 |
XM_005253898.1 |
3071 |
Missense Mutation |
GAG,GAT |
E758D |
XP_005253955.1 |
XM_006719445.2 |
3071 |
Missense Mutation |
GAG,GAT |
E758D |
XP_006719508.1 |
XM_011538437.1 |
3071 |
Missense Mutation |
GAG,GAT |
E758D |
XP_011536739.1 |
XM_011538439.1 |
3071 |
Missense Mutation |
GAG,GAT |
E749D |
XP_011536741.1 |
XM_017019408.1 |
3071 |
Missense Mutation |
GAG,GAT |
E758D |
XP_016874897.1 |
XM_017019409.1 |
3071 |
Missense Mutation |
GAG,GAT |
E758D |
XP_016874898.1 |
XM_017019410.1 |
3071 |
Missense Mutation |
GAG,GAT |
E758D |
XP_016874899.1 |
XM_017019411.1 |
3071 |
Missense Mutation |
GAG,GAT |
E749D |
XP_016874900.1 |
XM_017019412.1 |
3071 |
Missense Mutation |
GAG,GAT |
E588D |
XP_016874901.1 |
XM_017019413.1 |
3071 |
Missense Mutation |
GAG,GAT |
E579D |
XP_016874902.1 |
View Full Product Details