Product Details

SNP ID
rs149948309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56318433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGGTTTCCCCTTGAATCTTCAG[A/G]TCTGGGGTAAGTAAAGGTGGAATAG
Phenotype
MIM: 605861
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNPY2 PubMed Links

Gene Details

Gene
CNPY2
Gene Name
canopy FGF signaling regulator 2
There are no transcripts associated with this gene.

Gene
PAN2
Gene Name
PAN2 poly(A) specific ribonuclease subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127460.2 3755 Silent Mutation GAC,GAT D1126D NP_001120932.1
NM_001166279.1 3755 Silent Mutation GAC,GAT D1125D NP_001159751.1
NM_014871.4 3755 Silent Mutation GAC,GAT D1122D NP_055686.3

View Full Product Details