Product Details

SNP ID
rs140106457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:110523276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGAAATGTCAGAGTTGTAGGAC[A/G]TGGTAACAACCCAGGGTGCTGAAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB20 PubMed Links

Gene Details

Gene
RAB20
Gene Name
RAB20, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017817.2 1312 UTR 3 NP_060287.1

View Full Product Details