Product Details

SNP ID
rs141844727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:76880672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTTGTCACTAAATGGTTACTAA[C/T]GCTGTTTGAGTGGTCATAAATGAAT
Phenotype
MIM: 610521
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCTD12 PubMed Links

Gene Details

Gene
KCTD12
Gene Name
potassium channel tetramerization domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138444.3 5734 UTR 3 NP_612453.1

View Full Product Details