Product Details

SNP ID
rs149497796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:32504617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAATTCAACCTCTAAAATTCTG[C/T]CATCTGGCCCCTCAAAATTCATGTC
Phenotype
MIM: 615788
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MINOS1P1 PubMed Links

Gene Details

Gene
MINOS1P1
Gene Name
mitochondrial inner membrane organizing system 1 pseudogene 1
There are no transcripts associated with this gene.

Gene
N4BP2L2
Gene Name
NEDD4 binding protein 2 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278432.1 10312 Intron NP_001265361.1
NM_001320836.1 10312 Intron NP_001307765.1
NM_014887.2 10312 Intron NP_055702.1
NM_033111.4 10312 Intron NP_149102.3
XM_011534869.2 10312 Intron XP_011533171.1
XM_011534870.2 10312 Intron XP_011533172.1
XM_011534871.2 10312 Intron XP_011533173.1
XM_011534872.2 10312 Intron XP_011533174.1
XM_011534874.2 10312 Intron XP_011533176.1
XM_017020324.1 10312 Intron XP_016875813.1
XM_017020325.1 10312 Intron XP_016875814.1
XM_017020326.1 10312 Intron XP_016875815.1
XM_017020327.1 10312 Intron XP_016875816.1
XM_017020328.1 10312 Intron XP_016875817.1
XM_017020329.1 10312 Intron XP_016875818.1
XM_017020330.1 10312 Intron XP_016875819.1
XM_017020331.1 10312 Intron XP_016875820.1
XM_017020332.1 10312 Intron XP_016875821.1
XM_017020333.1 10312 Intron XP_016875822.1
XM_017020334.1 10312 Intron XP_016875823.1
XM_017020335.1 10312 Intron XP_016875824.1
XM_017020336.1 10312 Intron XP_016875825.1
XM_017020337.1 10312 Intron XP_016875826.1
XM_017020338.1 10312 Intron XP_016875827.1
XM_017020339.1 10312 Intron XP_016875828.1
XM_017020340.1 10312 Intron XP_016875829.1
XM_017020341.1 10312 Intron XP_016875830.1
XM_017020342.1 10312 UTR 3 XP_016875831.1
XM_017020343.1 10312 Intron XP_016875832.1
XM_017020344.1 10312 Intron XP_016875833.1
XM_017020345.1 10312 Intron XP_016875834.1
XM_017020346.1 10312 Intron XP_016875835.1
XM_017020347.1 10312 Intron XP_016875836.1
XM_017020348.1 10312 Intron XP_016875837.1
XM_017020349.1 10312 Intron XP_016875838.1
Gene
N4BP2L2-IT2
Gene Name
N4BPL2 intronic transcript 2
There are no transcripts associated with this gene.

View Full Product Details