Product Details

SNP ID
rs142172227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:34567765 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAACTCTTCTTGTCTTAAAATCT[A/G]TAAGTTCTGTGAAAAAGAAATTAGG
Phenotype
MIM: 606098
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNX6 PubMed Links

Gene Details

Gene
SNX6
Gene Name
sorting nexin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021249.3 1237 Missense Mutation ACA,ATA T247I NP_067072.3
NM_152233.2 1237 Missense Mutation ACA,ATA T375I NP_689419.2
XM_006720224.1 1237 Missense Mutation ACA,ATA T338I XP_006720287.1

View Full Product Details