Product Details

SNP ID
rs146989539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:67388233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGTTGTACCTTACCAGTGGGAA[C/T]GCCATTATCTTCCAGAGCAGACACG
Phenotype
MIM: 603907 MIM: 603930 MIM: 608007
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF2S1 PubMed Links

Gene Details

Gene
EIF2S1
Gene Name
eukaryotic translation initiation factor 2 subunit alpha
There are no transcripts associated with this gene.

Gene
GPHN
Gene Name
gephyrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024218.1 938 Intron NP_001019389.1
NM_020806.4 938 Intron NP_065857.1
XM_005267254.3 938 Intron XP_005267311.1
XM_011536340.2 938 Intron XP_011534642.1
XM_011536342.2 938 Intron XP_011534644.1
XM_011536343.2 938 Intron XP_011534645.1
XM_011536344.2 938 Intron XP_011534646.1
XM_011536345.2 938 Intron XP_011534647.1
XM_011536346.2 938 Intron XP_011534648.1
XM_011536347.2 938 Intron XP_011534649.1
XM_017020913.1 938 Intron XP_016876402.1
XM_017020914.1 938 Intron XP_016876403.1
XM_017020915.1 938 Intron XP_016876404.1
XM_017020916.1 938 Intron XP_016876405.1
XM_017020917.1 938 Intron XP_016876406.1
XM_017020918.1 938 Intron XP_016876407.1
XM_017020919.1 938 Intron XP_016876408.1
XM_017020920.1 938 Intron XP_016876409.1
XM_017020921.1 938 Intron XP_016876410.1
XM_017020922.1 938 Intron XP_016876411.1
XM_017020923.1 938 Intron XP_016876412.1
XM_017020924.1 938 Intron XP_016876413.1
XM_017020925.1 938 Intron XP_016876414.1
XM_017020926.1 938 Intron XP_016876415.1
Gene
PLEK2
Gene Name
pleckstrin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016445.2 938 Missense Mutation ATT,GTT I309V NP_057529.1
XM_011536644.2 938 Missense Mutation ATT,GTT I275V XP_011534946.1

View Full Product Details