Product Details

SNP ID
rs147705721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24144015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCCTTGGCCACAGCATCCCCA[C/T]GTTCTGAGAAGTACCTGCCAGAAGC
Phenotype
MIM: 600654 MIM: 602161 MIM: 612487
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMC9 PubMed Links

Gene Details

Gene
EMC9
Gene Name
ER membrane protein complex subunit 9
There are no transcripts associated with this gene.

Gene
MIR7703
Gene Name
microRNA 7703
There are no transcripts associated with this gene.

Gene
PSME1
Gene Name
proteasome activator subunit 1
There are no transcripts associated with this gene.

Gene
PSME2
Gene Name
proteasome activator subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002818.2 1371 Intron NP_002809.2
XM_006720213.1 1371 Missense Mutation CAT,CGT H94R XP_006720276.1
Gene
RNF31
Gene Name
ring finger protein 31
There are no transcripts associated with this gene.

View Full Product Details