Product Details

SNP ID
rs111381252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101304006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGATTTCTCTTTCAGAGTTGTA[C/G]ATTCAATAAACAACATCTGGGTTCT
Phenotype
MIM: 167405 MIM: 603521
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100507472 PubMed Links

Gene Details

Gene
LOC100507472
Gene Name
uncharacterized LOC100507472
There are no transcripts associated with this gene.

Gene
PCSK6
Gene Name
proprotein convertase subtilisin/kexin type 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291309.1 4437 UTR 3 NP_001278238.1
NM_002570.4 4437 UTR 3 NP_002561.1
NM_138319.3 4437 UTR 3 NP_612192.1
NM_138322.3 4437 Intron NP_612195.1
NM_138323.2 4437 Intron NP_612196.1
NM_138324.2 4437 Intron NP_612197.1
NM_138325.3 4437 Intron NP_612198.2
Gene
SNRPA1
Gene Name
small nuclear ribonucleoprotein polypeptide A'
There are no transcripts associated with this gene.

View Full Product Details