Product Details

SNP ID
rs116558850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43813058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCCATGCCTCATATTCCTCCTC[A/C]TCATTTTCATCATCAGTATTGAGTG
Phenotype
MIM: 600215
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MFAP1 PubMed Links

Gene Details

Gene
MFAP1
Gene Name
microfibrillar associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005926.2 1000 Missense Mutation GAG,GAT E272D NP_005917.2

View Full Product Details