Product Details
- SNP ID
-
rs138160608
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:64376809 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAATCTGATTCTCTTTTTCAGAA[A/T]CTTTAGGGGACAACCTAAAGAATTC
- Phenotype
-
MIM: 610696
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA0101
PubMed Links
Gene Details
- Gene
- KIAA0101
- Gene Name
- KIAA0101
View Full Product Details