Product Details

SNP ID
rs140030547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74212173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAGGATGAGTGGCGCTGGTCTC[A/G]GGATCCGCAGAGTCTACAGGGCCCT
Phenotype
MIM: 610745
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC33 PubMed Links

Gene Details

Gene
CCDC33
Gene Name
coiled-coil domain containing 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287181.1 569 Intron NP_001274110.1
NM_025055.4 569 Intron NP_079331.3
NM_182791.3 569 Intron NP_877592.2
XM_005254692.1 569 Intron XP_005254749.1
XM_011522085.2 569 Intron XP_011520387.1
XM_011522086.1 569 Intron XP_011520388.1
XM_011522087.2 569 Intron XP_011520389.1
XM_011522088.1 569 Intron XP_011520390.1
XM_011522089.2 569 Intron XP_011520391.1
XM_011522090.2 569 Intron XP_011520392.2
XM_017022623.1 569 Intron XP_016878112.1
XM_017022624.1 569 Intron XP_016878113.1
XM_017022625.1 569 Intron XP_016878114.1
XM_017022626.1 569 Intron XP_016878115.1
XM_017022627.1 569 Intron XP_016878116.1
XM_017022628.1 569 Intron XP_016878117.1
XM_017022629.1 569 UTR 5 XP_016878118.1
XM_017022630.1 569 Intron XP_016878119.1
XM_017022631.1 569 Intron XP_016878120.1
XM_017022632.1 569 Intron XP_016878121.1
XM_017022633.1 569 Intron XP_016878122.1
Gene
STRA6
Gene Name
stimulated by retinoic acid 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142617.1 569 Intron NP_001136089.1
NM_001142618.1 569 Intron NP_001136090.1
NM_001142619.1 569 Intron NP_001136091.1
NM_001142620.1 569 Intron NP_001136092.1
NM_001199040.1 569 Intron NP_001185969.1
NM_001199041.1 569 Intron NP_001185970.1
NM_001199042.1 569 Intron NP_001185971.1
NM_022369.3 569 Intron NP_071764.3
XM_011521883.1 569 UTR 5 XP_011520185.1
XM_011521884.1 569 Intron XP_011520186.1
XM_011521885.2 569 Intron XP_011520187.1
XM_017022478.1 569 Intron XP_016877967.1
XM_017022479.1 569 UTR 5 XP_016877968.1
XM_017022480.1 569 Intron XP_016877969.1

View Full Product Details