Product Details

SNP ID
rs143019993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:36809442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACTGAGTGTGCCAGAAATAAAC[C/T]TGGAGCAATTTTTAAATAAGCAAAA
Phenotype
MIM: 615626
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C15orf41 PubMed Links

Gene Details

Gene
C15orf41
Gene Name
chromosome 15 open reading frame 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130010.2 2111 UTR 3 NP_001123482.1
NM_001290232.1 2111 Intron NP_001277161.1
NM_001290233.1 2111 Intron NP_001277162.1
NM_001321756.1 2111 UTR 3 NP_001308685.1
NM_001321757.1 2111 UTR 3 NP_001308686.1
NM_001321758.1 2111 UTR 3 NP_001308687.1
NM_001321759.1 2111 UTR 3 NP_001308688.1
NM_001321760.1 2111 UTR 3 NP_001308689.1
NM_001321761.1 2111 UTR 3 NP_001308690.1
NM_032499.5 2111 UTR 3 NP_115888.1
XM_011522111.1 2111 Intron XP_011520413.1
XM_017022676.1 2111 UTR 3 XP_016878165.1
XM_017022677.1 2111 UTR 3 XP_016878166.1
Gene
CSNK1A1P1
Gene Name
casein kinase 1 alpha 1 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details