Product Details

SNP ID
rs138279409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28956003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATCTGGACAACTCTCCTCTGTCC[C/G]CACCTTCACCAAGGACCAAAAGCAG
Phenotype
MIM: 614525
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR4517 PubMed Links

Gene Details

Gene
MIR4517
Gene Name
microRNA 4517
There are no transcripts associated with this gene.

Gene
NFATC2IP
Gene Name
nuclear factor of activated T-cells 2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032815.3 619 Missense Mutation CCA,GCA P202A NP_116204.3

View Full Product Details