Product Details

SNP ID
rs139165037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66922277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGCGCACGACACCTTCAAACAG[C/T]GCCTGGACATTGTGGTGCAATGCCG
Phenotype
MIM: 603118 MIM: 614778 MIM: 179503
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDH16 PubMed Links

Gene Details

Gene
CDH16
Gene Name
cadherin 16
There are no transcripts associated with this gene.

Gene
FAM96B
Gene Name
family with sequence similarity 96 member B
There are no transcripts associated with this gene.

Gene
RRAD
Gene Name
RRAD, Ras related glycolysis inhibitor and calcium channel regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128850.1 869 Silent Mutation GCA,GCG A242A NP_001122322.1
NM_004165.2 869 Silent Mutation GCA,GCG A242A NP_004156.1

View Full Product Details